Product Details

SNP ID
rs13478114
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:17620854 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCGAGGTGCCTGGGCTCTGGA[G/T]TGAACTGCTCTGTGGAGGTCTTTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Sema3c PubMed Links

Gene Details

Gene
Sema3c
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013657.5 Intron NP_038685.3

View Full Product Details