Product Details
- SNP ID
-
rs13480601
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:44306419 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCCAGAGAGTGCTGAAGCAGCACC[G/T]GGCTGCTGAAGAGCGGGTGGGAAGG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Atg5
PubMed Links
Gene Details
- Gene
- Atg5
- Gene Name
- autophagy related 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_053069.5 |
|
Intron |
|
|
NP_444299.1 |
View Full Product Details