Product Details

SNP ID
rs4221957
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:129825026 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AGGTTCTGTACAAAGCGGGGTTCTC[T/C]TCTCTGAGCAGAGCTAACTGCCCTG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Gar1 PubMed Links

Gene Details

Gene
Gar1
Gene Name
GAR1 ribonucleoprotein homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_026578.3 1157 UTR 3 NP_080854.1
Gene
Rrh
Gene Name
retinal pigment epithelium derived rhodopsin homolog
There are no transcripts associated with this gene.

View Full Product Details