Product Details
- SNP ID
-
rs3023006
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:66841106 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTTCACTACAGAGACTTTATTC[C/A]TGGTGTAGCCATTGCTGCCAACATC
- Phenotype
-
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Tlr4
PubMed Links
Gene Details
- Gene
- Tlr4
- Gene Name
- toll-like receptor 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021297.2 |
2156 |
Missense Mutation |
|
|
NP_067272.1 |
View Full Product Details