Product Details

SNP ID
rs3720425
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:29229870 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AAACTTAGAGAGTGGCAATGGCAGT[A/G]AGATGCACTTCTGCTGCATTTATAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Cfap36 PubMed Links

Gene Details

Gene
Cfap36
Gene Name
cilia and flagella associated protein 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025740.3 Intron NP_080016.1
Gene
Smek2
Gene Name
SMEK homolog 2, suppressor of mek1 (Dictyostelium)
There are no transcripts associated with this gene.

View Full Product Details