Product Details

SNP ID
rs3090959
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:42627208 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTTATTATCTAAATTCTAGATGAT[A/G]CTAGAAAGCTCAATAGCATGGGCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Gabrb2 PubMed Links

Gene Details

Gene
Gabrb2
Gene Name
gamma-aminobutyric acid (GABA) A receptor, subunit beta 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008070.3 2169 UTR 3 NP_032096.1

View Full Product Details