Product Details

SNP ID
rs3023347
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:56710553 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTTCCTAATACAGTAATGTTTGTA[T/G]TTCTATGACCTTGTAAATATGGATG
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Pax9 PubMed Links

Gene Details

Gene
Pax9
Gene Name
paired box 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_011041.2 1897 UTR 3 NP_035171.1
Gene
Slc25a21
Gene Name
solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21
There are no transcripts associated with this gene.

View Full Product Details