Product Details

SNP ID
rs4231663
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.17:78377578 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCCAACTACAGCAGCATTTCACTG[G/A]CTGGGGACGGGAATGTAGACAACCT
Phenotype
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
Crim1 PubMed Links

Gene Details

Gene
Crim1
Gene Name
cysteine rich transmembrane BMP regulator 1 (chordin like)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015800.3 Intron NP_056615.1
Gene
Fez2
Gene Name
fasciculation and elongation protein zeta 2 (zygin II)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285940.1 Intron NP_001272869.1
NM_001285946.1 Intron NP_001272875.1
NM_001285949.1 Intron NP_001272878.1
NM_199448.3 Intron NP_955519.1

View Full Product Details