Product Details
- SNP ID
-
rs3675244
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:91060207 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTTCAAGCTAACAGCCTAGCACA[C/A]AAGCAAGAGAAAGGAAGGTGAAATT
- Phenotype
-
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Nrxn1
PubMed Links
Gene Details
- Gene
- Nrxn1
- Gene Name
- neurexin I
View Full Product Details