Product Details

SNP ID
rs3705122
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:23963350 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGTGTCTTGTAAATGATTAATAGCT[G/T]TGGTCATTAGTGCTTTTCCATGAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Zfp397 PubMed Links

Gene Details

Gene
Zfp397
Gene Name
zinc finger protein 397
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_027007.2 4166 UTR 3 NP_081283.2
Gene
Zscan30
Gene Name
zinc finger and SCAN domain containing 30
There are no transcripts associated with this gene.

View Full Product Details