Product Details
- SNP ID
-
rs13476200
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:158687382 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCAGAAAAGAGATGTTTTTTCA[C/T]AGGAGGAGTTTACCAGCTTAGAGTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Astn1
PubMed Links
Gene Details
- Gene
- Astn1
- Gene Name
- astrotactin 1
View Full Product Details