Product Details
- SNP ID
-
rs4223497
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:130391030 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGAGTAGGAAATTGCAGGGAACAGG[A/G]CCCTCTTCAAAGGTGACCTGACAGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cpxm1
PubMed Links
Gene Details
- Gene
- Cpxm1
- Gene Name
- carboxypeptidase X 1 (M14 family)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019696.2 |
2166 |
Silent Mutation |
|
|
NP_062670.2 |
View Full Product Details