Product Details

SNP ID
rs4230877
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.15:79915806 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACAAACAACCAGACACACAGAAGT[C/T]TCTAAAGACATTTTAATGTATTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Apobec3 PubMed Links

Gene Details

Gene
Apobec3
Gene Name
apolipoprotein B mRNA editing enzyme, catalytic polypeptide 3
There are no transcripts associated with this gene.

Gene
Cbx7
Gene Name
chromobox 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144811.3 Intron NP_659060.1

View Full Product Details