Product Details
- SNP ID
-
rs4231296
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:87730533 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTAGACTGTTAGGAACTTTCTG[A/T]CCCGTGGAGGAGGAGCTGCCCCGGA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Bach1
PubMed Links
Gene Details
- Gene
- Bach1
- Gene Name
- BTB and CNC homology 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007520.2 |
3045 |
UTR 3 |
|
|
NP_031546.1 |
View Full Product Details