Product Details

SNP ID
rs3658371
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.18:14977931 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
AACACTTACAGGTTTGGTAGCTTTG[A/G]CTGGCAATATTTATTAGAAAGACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Kctd1 PubMed Links

Gene Details

Gene
Kctd1
Gene Name
potassium channel tetramerisation domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142731.1 Intron NP_001136203.1
NM_134112.5 Intron NP_598873.2

View Full Product Details