Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCKSL1
Entrez Gene ID
102124661
Gene Name
MARCKS like 1
Gene Aliases
-
Chromosome Location
Chr.1: 195648715 - 195651096 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544104.1 XP_005544161.1 1 - 2 298 144

View Full Product Details