Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYSMD2
Entrez Gene ID
102121018
Gene Name
LysM domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 28711293 - 28740372 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005559567.2 XP_005559624.1 2 - 3 634 72
XM_015452831.1 XP_015308317.1 2 - 3 663 72

View Full Product Details