Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMGA2
Entrez Gene ID
102118074
Gene Name
high mobility group AT-hook 2
Gene Aliases
-
Chromosome Location
Chr.11: 64897660 - 65037320 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431160.1 XP_015286646.1 4 - 5 1341 80

View Full Product Details