Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFP90
Entrez Gene ID
102140885
Gene Name
ZFP90 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.20: 56773212 - 56810930 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592354.2 XP_005592411.1 3 - 4 452 70
XM_005592355.2 XP_005592412.1 3 - 4 442 70
XM_005592356.2 XP_005592413.2 2 - 3 477 70
XM_015443072.1 XP_015298558.1 1 - 2 882 70
XM_015443073.1 XP_015298559.1 3 - 4 382 70
XM_015443074.1 XP_015298560.1 3 - 4 354 70
XM_015443075.1 XP_015298561.1 3 - 4 452 70
XM_015443076.1 XP_015298562.1 3 - 4 442 70
XM_015443077.1 XP_015298563.1 3 - 4 452 70

View Full Product Details