Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RABIF
Entrez Gene ID
102130384
Gene Name
RAB interacting factor
Gene Aliases
-
Chromosome Location
Chr.1: 61266583 - 61278463 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005540463.2 XP_005540520.1 1 - 2 1926 63

View Full Product Details