Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTF3
Entrez Gene ID
102128547
Gene Name
neurotrophin 3
Gene Aliases
-
Chromosome Location
Chr.11: 5649007 - 5713195 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005569876.2 XP_005569933.1 1 - 2 220 97

View Full Product Details