Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIKESHI
Entrez Gene ID
102126161
Gene Name
heat shock protein nuclear import factor hikeshi
Gene Aliases
-
Chromosome Location
Chr.14: 81878234 - 81919208 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579283.2 XP_005579340.1 4 - 5 774 133
XM_015435583.1 XP_015291069.1 5 - 6 781 133
XM_015435584.1 XP_015291070.1 5 - 6 914 133

View Full Product Details