Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARMCX2
Entrez Gene ID
102130211
Gene Name
armadillo repeat containing X-linked 2
Gene Aliases
-
Chromosome Location
Chr.X: 98792158 - 98796762 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005594159.1 XP_005594216.1 6 - 6 2088 154
XM_005594160.2 XP_005594217.1 5 - 5 1992 154
XM_015444300.1 XP_015299786.1 6 - 6 2100 154
XM_015444301.1 XP_015299787.1 6 - 6 2104 154
XM_015444302.1 XP_015299788.1 6 - 6 2111 154
XM_015444303.1 XP_015299789.1 6 - 6 2056 154
XM_015444304.1 XP_015299790.1 5 - 5 1996 154
XM_015444305.1 XP_015299791.1 6 - 6 2044 154
XM_015444306.1 XP_015299792.1 6 - 6 2067 154
XM_015444307.1 XP_015299793.1 5 - 5 2003 154
XM_015444308.1 XP_015299794.1 5 - 5 1980 154

View Full Product Details