Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCHR1
Entrez Gene ID
102137192
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
-
Chromosome Location
Chr.10: 10457556 - 10462256 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005567181.2 XP_005567238.1 1 - 2 1615 84

View Full Product Details