Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCFC1R1
Entrez Gene ID
102142598
Gene Name
host cell factor C1 regulator 1
Gene Aliases
-
Chromosome Location
Chr.20: 3300368 - 3302027 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005591029.1 XP_005591086.1 1 - 2 444 131

View Full Product Details