Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
102144723
Gene Name
exosome component 4
Gene Aliases
-
Chromosome Location
Chr.8: 145715016 - 145717046 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005564313.2 XP_005564370.1 2 - 3 506 65

View Full Product Details