Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD6
Entrez Gene ID
102145682
Gene Name
caspase recruitment domain family member 6
Gene Aliases
-
Chromosome Location
Chr.6: 41321629 - 41335649 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005556789.2 XP_005556846.1 1 - 2 462 85

View Full Product Details