Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG14
Entrez Gene ID
102126813
Gene Name
ALG14 UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
-
Chromosome Location
Chr.1: 131840177 - 131933700 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005542655.2 XP_005542712.1 1 - 2 191 65
XM_005542656.2 XP_005542713.1 1 - 2 198 65
XM_015432549.1 XP_015288035.1 1 - 2 198 65

View Full Product Details