Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNG11
Entrez Gene ID
102133905
Gene Name
G protein subunit gamma 11
Gene Aliases
-
Chromosome Location
Chr.3: 118054630 - 118059450 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005550207.2 XP_005550264.1 1 - 2 522 158

View Full Product Details