Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP20
Entrez Gene ID
102144643
Gene Name
cilia and flagella associated protein 20
Gene Aliases
C20H16orf80
Chromosome Location
Chr.20: 46160332 - 46176505 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592094.2 XP_005592151.1 5 - 6 901 78

View Full Product Details