Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MFAP3
Entrez Gene ID
102127417
Gene Name
microfibril associated protein 3
Gene Aliases
-
Chromosome Location
Chr.6: 153299326 - 153318138 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005558343.2 XP_005558400.1 2 - 3 1681 100
XM_005558344.2 XP_005558401.1 2 - 3 496 100
XM_005558345.2 XP_005558402.1 2 - 3 3164 100
XM_005558346.2 XP_005558403.1 3 - 4 678 100
XM_005558347.2 XP_005558404.1 2 - 3 482 100
XM_015452169.1 XP_015307655.1 3 - 4 657 100
XM_015452170.1 XP_015307656.1 3 - 4 808 100
XM_015452171.1 XP_015307657.1 3 - 4 1737 100
XM_015452172.1 XP_015307658.1 2 - 3 481 100
XM_015452173.1 XP_015307659.1 2 - 3 647 100

View Full Product Details