Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZNRD2
Entrez Gene ID
102134846
Gene Name
zinc ribbon domain containing 2
Gene Aliases
SSSCA1
Chromosome Location
Chr.14: 8917214 - 8918836 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005577268.2 XP_005577325.1 2 - 3 517 82

View Full Product Details