Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D1B
Entrez Gene ID
102132319
Gene Name
SH2 domain containing 1B
Gene Aliases
-
Chromosome Location
Chr.1: 89316508 - 89335708 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015454409.1 XP_015309895.1 2 - 3 322 97

View Full Product Details