Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MED18
Entrez Gene ID
102138806
Gene Name
mediator complex subunit 18
Gene Aliases
-
Chromosome Location
Chr.1: 199868459 - 199875500 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005544236.2 XP_005544293.1 1 - 2 206 115

View Full Product Details