Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PRMT6
Entrez Gene ID
102117289
Gene Name
protein arginine methyltransferase 6
Gene Aliases
-
Chromosome Location
Chr.1: 119851656 - 119858653 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015431982.1 XP_015287468.1 1 - 1 526 75
XM_015431988.1 XP_015287474.1 1 - 1 526 75

View Full Product Details