Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH20
Entrez Gene ID
102138455
Gene Name
protocadherin 20
Gene Aliases
-
Chromosome Location
Chr.17: 41093542 - 41113286 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585952.2 XP_005586009.1 4 - 5 2246 110

View Full Product Details