Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AGTR1
Entrez Gene ID
102130940
Gene Name
angiotensin II receptor type 1
Gene Aliases
-
Chromosome Location
Chr.2: 50903128 - 50948092 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005546050.2 XP_005546107.1 1 - 1 131 82

View Full Product Details