Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEX3B
Entrez Gene ID
102136526
Gene Name
mex-3 RNA binding family member B
Gene Aliases
-
Chromosome Location
Chr.7: 60540409 - 60544741 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005560277.1 XP_005560334.1 1 - 2 680 97

View Full Product Details