Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAAR6
Entrez Gene ID
102136851
Gene Name
trace amine associated receptor 6
Gene Aliases
-
Chromosome Location
Chr.4: 36720144 - 36721181 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005551853.1 XP_005551910.1 1 - 1 587 130

View Full Product Details