Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PATE2
Entrez Gene ID
102140295
Gene Name
prostate and testis expressed 2
Gene Aliases
-
Chromosome Location
Chr.14: 121596148 - 121602993 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005580092.2 XP_005580149.1 3 - 4 2553 163

View Full Product Details