Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIXL1
Entrez Gene ID
102147071
Gene Name
Mix paired-like homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 85063148 - 85066724 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005541009.2 XP_005541066.1 1 - 2 568 144

View Full Product Details