Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNLRB2
Entrez Gene ID
102143645
Gene Name
dynein light chain roadblock-type 2
Gene Aliases
-
Chromosome Location
Chr.20: 68796499 - 68806370 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005592622.2 XP_005592679.1 1 - 2 123 122

View Full Product Details