Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NCR2
Entrez Gene ID
102129442
Gene Name
natural cytotoxicity triggering receptor 2
Gene Aliases
NKp44
Chromosome Location
Chr.4: 129334628 - 129350205 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319537.1 NP_001306466.1 3 - 4 612 167
GenBank mRNA AM404415.1 - 3 - 4 612 167

View Full Product Details