Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
102127340
Gene Name
FA core complex associated protein 100
Gene Aliases
C16H17orf70
Chromosome Location
Chr.16: 79155979 - 79169521 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005585240.2 XP_005585297.1 4 - 5 1471 78
XM_005585241.2 XP_005585298.1 4 - 5 1663 78
XM_015438956.1 XP_015294442.1 4 - 5 1470 78
XM_015438957.1 XP_015294443.1 4 - 5 1342 78
XM_015438958.1 XP_015294444.1 3 - 4 1223 78

View Full Product Details