Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A4
Entrez Gene ID
102137112
Gene Name
solute carrier family 10 member 4
Gene Aliases
-
Chromosome Location
Chr.5: 48580410 - 48586203 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005554850.2 XP_005554907.1 2 - 3 1015 93

View Full Product Details