Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOXD13
Entrez Gene ID
102126260
Gene Name
homeobox D13
Gene Aliases
-
Chromosome Location
Chr.12: 65056212 - 65059189 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005573548.1 XP_005573605.1 1 - 2 712 114

View Full Product Details