Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPBG
Entrez Gene ID
102132149
Gene Name
trophoblast glycoprotein
Gene Aliases
WAIF1
Chromosome Location
Chr.4: 91496669 - 91505156 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005552483.2 XP_005552540.1 2 - 3 1523 134
XM_015449153.1 XP_015304639.1 1 - 2 1629 134

View Full Product Details