Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB25
Entrez Gene ID
102147199
Gene Name
zinc finger and BTB domain containing 25
Gene Aliases
-
Chromosome Location
Chr.7: 128827094 - 128849395 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005561469.2 XP_005561526.1 1 - 2 180 70

View Full Product Details