Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRSS23
Entrez Gene ID
102131149
Gene Name
serine protease 23
Gene Aliases
-
Chromosome Location
Chr.14: 82354743 - 82375182 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005579295.2 XP_005579352.1 1 - 2 195 100

View Full Product Details