Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BMP10
Entrez Gene ID
102133809
Gene Name
bone morphogenetic protein 10
Gene Aliases
-
Chromosome Location
Chr.13: 41117934 - 41124786 on Build M_fas_5.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005575695.1 XP_005575752.1 1 - 2 499 120

View Full Product Details